Skip to Content
Merck
All Photos(1)

Key Documents

8.14566

Sigma-Aldrich

Diisopropylazodicarboxylate

for synthesis

Synonym(s):

Diisopropylazodicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHO2CN=NCO2CH(CH3)2
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
219-502-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.04 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

potency

2150 mg/kg LD50, skin (Rabbit)

bp

75 °C/33.3 Pa

transition temp

flash point 106 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(=NC(=O)OC(C)C)C(=O)OC(C)C

InChI

1S/C8H14N2O4/c1-5(2)13-7(11)9-10-8(12)14-6(3)4/h5-6H,1-4H3

InChI key

VVWRJUBEIPHGQF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.038 - 1.048
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

222.8 °F

Flash Point(C)

106 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service