Skip to Content
Merck
All Photos(1)

Key Documents

8.11866

Sigma-Aldrich

Crown ether/Dicyclohexyl-18-crown-6

(mixture of cis- and trans-isomers) for synthesis

Synonym(s):

Crown ether/Dicyclohexyl-18-crown-6, 2,5,8,15,18,21-Hexaoxatricyclo[20.4.0.09.14] hexacosane, Dicyclohexyl-18-crown-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36O6
CAS Number:
Molecular Weight:
372.50
MDL number:
UNSPSC Code:
12352005
EC Index Number:
240-216-4
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (GC)

form

powder

potency

176 mg/kg LD50, oral (Rat)

mp

46-50 °C

transition temp

flash point 113 °C

storage temp.

2-30°C

SMILES string

O1C2C(OCCOCCOC3C(OCCOCC1)CCCC3)CCCC2

InChI

1S/C20H36O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h17-20H,1-16H2

InChI key

BBGKDYHZQOSNMU-UHFFFAOYSA-N

Application

  • Complexes formation and thermodynamic parameters between 15-crown-5, 1, 4, 8, 11-tetrathiacyclotetradecane and 1, 7-diaza-12-crown-4 macrocyclic ligands: This study includes details on Dicyclohexyl-18-crown-6 (DCH18C6) and its interactions with other crown ethers and their solvation properties in various solvents. It highlights the thermodynamic behaviors of these macrocyclic compounds in solution (MS Kazemi, B Shamshirgaran, 2016).
  • Silver (I) extraction with benzo-18-crown-6 ether from water into 1, 2-dichloroethane: This research explores the extraction characteristics of silver ions using benzo-18-crown-6 ether which is similar in behavior to Dicyclohexyl-18-crown-6, thus providing comparative insights into the use of crown ethers for ion transport and extraction (Y Kudo, S Ikeda, S Morioka, S Tomokata, 2017).
  • Separation and enrichment of Rb (I) in dicyclohexano 18 crown 6 (DCH18C6)/PVC polymer inclusion membrane assisted by electric field: This article discusses the application of DCH18C6 in polymer inclusion membranes for the selective separation and enrichment of rubidium ions, showcasing the utility of crown ethers in membrane technology (X Meng, Y Tian, MY Tang, R Qiao, 2021).
  • Extraction of Gd3+ and UO22+ Ions Using Polystyrene Grafted Dibenzo Crown Ether (DB18C6) with Octanol and Nitrobenzene: This molecular dynamics study provides insights into the extraction processes of metal ions using a structurally similar crown ether, which could reflect on the extraction capabilities of Dicyclohexyl-18-crown-6 in similar settings (P Sappidi, S Namsani, SM Ali, JK Singh, 2018).
  • A facile preparation of dicyclohexano-18-crown-6 ether impregnated titanate nanotubes for strontium removal from acidic solution: Demonstrates the use of DCH18C6 for the removal of strontium from acidic solutions, emphasizing the crown ether′s potential in environmental cleanup and radioactive waste management (J Ma, Y Zhang, J Ouyang, X Wu, J Luo, S Liu, 2019).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service