Skip to Content
Merck
All Photos(1)

Key Documents

8.08504

Sigma-Aldrich

Pentanal

for synthesis

Synonym(s):

Pentanal, Valeraldehyde, Amylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CHO
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352114
EC Index Number:
203-784-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

36 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

205 °C

potency

3200 mg/kg LD50, oral (Rat)
4850 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-6.8 % (v/v)

mp

-91 °C

transition temp

flash point 7 °C

solubility

11.7 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O=CCCCC

InChI

1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3

InChI key

HGBOYTHUEUWSSQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.808 - 0.815
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service