Skip to Content
Merck
All Photos(2)

Key Documents

79102BC

Millipore

GDS PPMX Diamond Tips

BioControl, For use with GDS PickPen® PIPETMAX®, capacity 200 μL, filter, sterile, Blister Refill

Synonym(s):

Sterile Pipette Tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121610

description

For use with GDS PICKPEN™ PIPETMAX™ System

sterility

sterile

feature

filter
Blister Refill

manufacturer/tradename

BioControl Systems 79102

storage condition

room temperature

technique(s)

PCR: suitable

capacity

200 μL

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PICKPEN® PIPETMAX® System

storage temp.

room temp

General description

GDS PPMX Diamond Tips are the consumable accessories of the GDS PickPen® PIPETMAX® (PPMX) device. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. As part of the automated sample preparation for Assurance® GDS assays, the PPMX was designed to complete reagent and sample preparation for the GDS PCR.

Application

The Assurance® GDS PPMX Diamond Tips are used with the PickPen® PIPETMAX® device to transfer liquids during automated Assurance® GDS sample preparation.

Components

10 Blister refills of 96 racked tips

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service