Skip to Content
Merck
All Photos(1)

Key Documents

444215

Sigma-Aldrich

MMP-2/MMP-9 Substrate I, Fluorogenic

A fluorogenic substrate for both the 72 kDa human fibroblast gelatinase (HFG; MMP-2) and the 92 kDa human neutrophil gelatinase (HNG; MMP-9).

Synonym(s):

MMP-2/MMP-9 Substrate I, Fluorogenic, Gelatinase A/Gelatinase B Substrate I, DNP-PLGMWSR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H61N13O13S
Molecular Weight:
1012.10
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: soluble

fluorescence

λex ~280 nm
λem ~360 nm

shipped in

ambient

storage temp.

−20°C

General description

A fluorogenic substrate for both the 72 kDa human fibroblast gelatinase (HFG; MMP-2) and the 92 kDa human neutrophil gelatinase (HNG; MMP-9) (excitation maximum: 280 nm; emission maximum: 360 nm).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A fluorogenic substrate for both human fibroblast gelatinase (HFG; MMP-2) and human neutrophil gelatinase (HNG; MMP-9).
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

DNP-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to months at -20°C.

Other Notes

Netzel-Arnett, S., et al. 1991. Anal. Biochem. 195, 86.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service