Skip to Content
Merck
All Photos(1)

Key Documents

238811

Sigma-Aldrich

Cdk9 Inhibitor II

The Cdk9 Inhibitor II, also referenced under CAS 140651-18-9, controls the biological activity of Cdk9. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk9 Inhibitor II, 4-(3,5-Diamino-1H-pyrazol-4-ylazo)-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N6O
CAS Number:
Molecular Weight:
218.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1nc(c(c1N)N=Nc2ccc(cc2)O)N

InChI

1S/C9H10N6O/c10-8-7(9(11)15-14-8)13-12-5-1-3-6(16)4-2-5/h1-4,16H,(H5,10,11,14,15)

InChI key

AYZRKFOEZQBUEA-UHFFFAOYSA-N

General description

A cell-permeable azo-pyrazole compound that acts as a potent, ATP-competitive, and Cdk9-selective inhibitor (IC50 = 350 nM, using Cdk9/T1). It inhibits Cdk1/B, Cdk2/E, Cdk2/A, Cdk4/D1, Cdk7/H, and p70S6K only at much higher concentrations (IC50 = 44, 20, 69, 13.5, 26, and 10 µM, respectively) and shows little or no activity against eight other commonly studied kinases even at concentrations as high as 10 µM. Shown to exhibit antiproliferative activity in human tumor cell lines.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk9
Product competes with ATP.
Reversible: no
Target IC50: 350 nM, using Cdk9/T1

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Krystof, V., et al. 2006. J. Med. Chem.49, 6500.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XinTao Gao et al.
Molecular oncology, 15(7), 1901-1920 (2021-05-02)
Androgen receptor (AR) is the principal molecule in prostate cancer (PCa) etiology and therapy. AR re-activation still remains a major challenge during treatment of castration-resistant prostate cancer (CRPC) tumors that relapse after castration therapies. Recent reports have indicated the enrichment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service