Skip to Content
Merck
All Photos(1)

Documents

12-510

Sigma-Aldrich

Goat Anti-Rabbit IgG (H+L) Antibody, Rhodamine conjugate

Upstate®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

goat

Quality Level

conjugate

TRITC conjugate
rhodamine conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Goat anti-Rabbit IgG (H+L) Antibody, Rhodamine conjugate detects level of Rabbit IgG (H+L) & has been published & validated for use in FC, IF, IC, IH.

Quality

Routinely evaluated by immunomicroscopy, flow cytometry or FACS analysis, as well as other antibody based fluorescent assays requiring lot-to-lot consistency

Storage and Stability

Lyophilized: Store at 4 °C prior to restoration. Restore with 0.25 mL of deionized water (or equivalent). This product is stable for several weeks at 4 °C as an undiluted liquid. For extended storage mix product with glycerol to 50% and then aliquot contents and freeze at -20 °C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. Dilute only prior to immediate use. Expiration date is one year from date of restoration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service