Skip to Content
Merck
All Photos(3)

Key Documents

06-1077-I

Sigma-Aldrich

Anti-IL-22RA-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-22 receptor subunit alpha-1, CRF2-9, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, ZcytoR11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... IL22RA1(58985)

General description

Interleukin-22 receptor subunit alpha-1 (UniProt Q8N6P7; also known as CRF2-9, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, ZcytoR11) is encoded by the IL22RA1 (also known as IL22R) gene (Gene ID 58985) in human. IL-22RA-1 is a member of the type II cytokine receptor family that complexes with IL-10RB and mediates JAK/STAT signaling. IL22 binding to heterodimer is greater than binding to IL22RA1 alone. IL-22RA-1 is expressed in colon, liver, lung, pancreas and kidney, however, no expression is observed in immune cells, such as monocytes, T-cells, and NK-cells. It is also detected in normal blood brain barrier endothelial cells as well as in multiple sclerosis lesions. Higher expression has been observed in synovial fluid cells from rheumatoid arthritis and spondyloarthropathy patients.

Specificity

This polyclonal antibody targets a C-terminal sequence within the cytoplasmic domain of human IL-22 receptor subunit alpha-1.

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the C-terminal region of human IL-22RA-1 cytoplasmic domain.

Application

Flow Cytometry Analysis: 1 µg from a representative lot detected IL-22RA-1 immunoreactivity among one million 4% paraformaldehyde-fixed, 0.2% Triton X-100-permeabilized SW480 human colon cancer cells.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected IL-22RA-1 in human tonsil and prostate tissue sections.
Research Category
Inflammation & Immunology
This rabbit polyclonal Anti-IL-22RA-1 Antibody, Cat. No. 06-1077-I is validated for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting for the detection of Interleukin-22 receptor subunit alpha-1.

Quality

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected IL-22RA-1 in 10 µg of mouse spleen tissue lysate.

Target description

~63 kDa observed. 61.54 kDa (signal peptide a.a. 1-15 removed) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 06-1077

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service