Z742941
Julabo® hightech bridge mounted circulator
Model SE-Z, temperature 20-300 °C, AC/DC input 230 V AC, US 3-pin plug (6-20P)
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9252218
temperature measuring range
20-300 °C
General description
JULABO bridge mounted circulator with extendable stainless steel bridge. The adjustable bridge permits use of the circulator with a wide variety of bath tanks.
This model is suitable for both internal and external temperature control tasks. It has the power to control the temperature of even large bath tanks up to 100 L.
An integrated cooling coil enables temperature control below or near the ambient temperature.
This model is suitable for both internal and external temperature control tasks. It has the power to control the temperature of even large bath tanks up to 100 L.
An integrated cooling coil enables temperature control below or near the ambient temperature.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)