Key Documents
Z742352
2MAG Stirring bar remover PTFE
flexible with extra strong permanent magnet
Synonym(s):
magnetic extraction tool
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
manufacturer/tradename
2mag 45070
General description
PTFE-coated flexible stirring bar remover with extremely strong permanent neodym magnet, length 70 cm. Universal chemical resistance also in aggressive media, ideal for fast removal of heavy and big magnetic stirring bars with a weight up to 400 g.
Choose from one of the most recent versions:
Certificates of Analysis (COA)