Key Documents
Z615692
Julabo® bath test tube racks
to fit 90 microtiter tubes, for water baths, diam. 11-12 mm
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
Pricing and availability is not currently available.
material
polypropylene
feature
Holds 90 x 11-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970382
parameter
+80 °C temp. limit
diam.
11-12 mm
General description
Polypropylene tray and test tube inserts can be used up to +80 °C. When tempering solvent-containing or acidic samples above 60 °C, it is recommended using stainless steel test tube inserts, as the plastic can be attacked.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)