Skip to Content
Merck
All Photos(1)

Key Documents

Z406430

Wood applicators

useful for stirring, sample transfer, and application of grease to stopcocks, pkg of 864 ea

Synonym(s):

Wooden sticks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.35

packaging

pkg of 864 ea

General description

Smooth white birch sticks are useful for stirring, sample transfer, and application of grease to stopcocks.

Each applicator is 5 3/4 in. L × 7/8 in. diam.

Application

Wood applicators have been used to weigh fecalsamples.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High?Yield Purification of Giardia intestinalis Cysts from Fecal Samples
Paul O, et al.
Current Protocols in Microbiology, 59(1) (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service