Skip to Content
Merck
All Photos(1)

Documents

W202207

Sigma-Aldrich

Allyl cinnamate

≥99%

Synonym(s):

Allyl 3-phenylacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCO2CH2CH=CH2
CAS Number:
Molecular Weight:
188.22
FEMA Number:
2022
EC Number:
Council of Europe no.:
334
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.741
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.566 (lit.)

bp

150-152 °C/15 mmHg (lit.)

density

1.053 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic; spicy; fruity; pineapple

SMILES string

C=CCOC(=O)\C=C\c1ccccc1

InChI

1S/C12H12O2/c1-2-10-14-12(13)9-8-11-6-4-3-5-7-11/h2-9H,1,10H2/b9-8+

InChI key

KCMITHMNVLRGJU-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Acaricidal potential of active components derived from Alpinia galanga rhizome oils and their derivatives against Haemaphysalis longicornis (Acari: Ixodidae).: This research explores the acaricidal activity of allyl cinnamate, a component derived from Alpinia galanga, highlighting its potential as a natural pesticide. The findings could lead to eco-friendly solutions in pest management, particularly in agriculture and livestock protection (Kang et al., 2022).

  • Biocide Potentiation Using Cinnamic Phytochemicals and Derivatives.: Investigates the synergistic effects of allyl cinnamate with other biocides, demonstrating its potential to enhance antimicrobial efficacy against various pathogens. This study underscores the role of cinnamate derivatives in developing advanced antimicrobial strategies, critical for both healthcare and industry applications (Malheiro et al., 2019).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service