Skip to Content
Merck
All Photos(1)

Documents

T79758

Sigma-Aldrich

Trimethyl phosphonoacetate

98%

Synonym(s):

(Carboxymethyl)phosphonic acid trimethyl ester, NSC 84262

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)CH2CO2CH3
CAS Number:
Molecular Weight:
182.11
Beilstein:
1865177
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.437 (lit.)

bp

118 °C/0.85 mmHg (lit.)
265-268 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CP(=O)(OC)OC

InChI

1S/C5H11O5P/c1-8-5(6)4-11(7,9-2)10-3/h4H2,1-3H3

InChI key

SIGOIUCRXKUEIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Intramolecular Mannich-type reactions to produce the sarain A diazatricyclic core
  • Organocatalytic oxa-Michael reactions
  • Prenylation and geranylation of oxindoles
  • Intramolecular Horner-Wadsworth-Emmons reactions
  • Olefin cross-metathesis / heterocyclization

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

B R DeMark et al.
Archives of biochemistry and biophysics, 223(2), 360-368 (1983-06-01)
The effects of the inhibitors trimethylacetyl phosphate and cAMP have been determined in reactions catalyzed by D-glyceraldehyde-3-phosphate dehydrogenase. These inhibitors must influence the oxidation of aldehydes through substrate dependent cooperative conformational changes. Both trimethylacetyl phosphate and cAMP give sigmoidal 1/V
Alastair B Ross et al.
The Journal of nutrition, 143(6), 766-773 (2013-04-26)
Epidemiological studies consistently find that diets rich in whole-grain (WG) cereals lead to decreased risk of disease compared with refined grain (RG)-based diets. Aside from a greater amount of fiber and micronutrients, possible mechanisms for why WGs may be beneficial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service