Key Documents
SYNA381410
Synthware vacuum adapter with PTFE stopcock
joint: ST/NS 14/10
Synonym(s):
Synthware A381410, glass adapter with stopcock
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/10
packaging
pack of 1 ea
manufacturer/tradename
A381410
Related Categories
General description
Synthware vacuum adapter with PTFE stopcock for microscale applications. Hose connection is 8 mm O.D. and joint is 14/10. Stopcock bore is 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)