Key Documents
SYNA351922
Synthware distillation adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 19/22, side joint: 19/22, side arm angle: 75 deg
Synonym(s):
Synthware A351922, vacuum adapter, distillation adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
bottom joint: ST/NS19/22
side joint: 19/22
joint: ST/NS top19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A351922
General description
For use as a component in atmospheric or vacuum distillations.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)