GF25606368
Brass alloy, Cu63Zn37
200mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter, as drawn
Synonym(s):
CU027200
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
30102010
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 256-063-68
L × wall thickness
200 mm × 0.25 mm
O.D. × I.D.
1.5 mm × 1 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service