DWKW242824-A
Wheaton® Dropping Bottle
natural bottle, capacity 10 mL, case of 1000
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
natural bottle
packaging
pkg of 1 packs
case of 1000
manufacturer/tradename
WHEATON® W242824-A
capacity
10 mL
Related Categories
Application
Opthalmic, IVD Kits, Other
Features and Benefits
- Bottle made of natural or white LDPE (Low Density Polyethylene
- White LDPE bottle helps protect UV light sensitive samples
- Dropper tip caps and tips sold separately
- Packaged in cases of 100 or 1000
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)