CLS6795170
Corning® Scholar 170 hot plate
AC/DC input 120 V AC, US 3-pin plug, plate W × L 5 in. × 5 in.
Synonym(s):
stirrers
About This Item
Recommended Products
material
enameled steel plate
sterility
non-sterile
manufacturer/tradename
Corning 6795-170
availability
not available in EU
parameter
25 °C temp. range (360 °C)
plate W × L
5 in. × 5 in.
color
none
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service