Skip to Content
Merck
All Photos(1)

Key Documents

900533

Sigma-Aldrich

(t-Bu)PhCPhos Pd G4

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H48N3O3PPdS
CAS Number:
Molecular Weight:
788.29
UNSPSC Code:
12161600
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder or crystals

feature

generation 4

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH]1C)[P](C=4C=CC=CC4)(C5=CC=CC=C5C=6C(=CC=CC6N(C)C)N(C)C)C(C)(C)C)C

InChI

InChI=1S/C26H33N2P.C13H12N.CH4O3S.Pd/c1-26(2,3)29(20-14-9-8-10-15-20)24-19-12-11-16-21(24)25-22(27(4)5)17-13-18-23(25)28(6)7;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h8-19H,1-7H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;-1;;+2/p-1

Application

A fourth generation (G4) Buchwald precatalyst that is similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations found when using the third generation precatalysts. It is air, moisture, and thermally-stable and shows good solubility in common organic solvents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Angewandte Chemie (International ed. in English), 54(28), 8259-8262 (2015-06-03)
In Pd-catalyzed C-N cross-coupling reactions, α-branched secondary amines are difficult coupling partners and the desired products are often produced in low yields. In order to provide a robust method for accessing N-aryl α-branched tertiary amines, new catalysts have been designed

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service