Skip to Content
Merck
All Photos(1)

Key Documents

731986

Sigma-Aldrich

N-Ethyl-d5-ethanolamine

98 atom % D, 97% (CP)

Synonym(s):

(2-Ethyl-d5-amino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2D5NHCH2CH2OH
Molecular Weight:
94.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

bp

169-170 °C (lit.)

density

0.964 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])NCCO

InChI

1S/C4H11NO/c1-2-5-3-4-6/h5-6H,2-4H2,1H3/i1D3,2D2

InChI key

MIJDSYMOBYNHOT-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service