Skip to Content
Merck
All Photos(3)

Key Documents

657123

Sigma-Aldrich

4-Amino-3-iodobenzonitrile

98%

Synonym(s):

1-Amino-2-iodo-4-cyanobenzene, 4-Cyano-2-iodoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(NH2)CN
CAS Number:
Molecular Weight:
244.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

112-115 °C (lit.)

SMILES string

Nc1ccc(cc1I)C#N

InChI

1S/C7H5IN2/c8-6-3-5(4-9)1-2-7(6)10/h1-3H,10H2

InChI key

UOWVTQFTEAYDLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Hydroamination-double hydroarylation for synthesis of fuzed carbazoles
  • Cycloisomerizations of aromatic homo- and bis-homopropargylic amine / amide intermediates
  • Asymmetric organocatalytic condensation and cycloaddition
  • Regioselective aryl C-H bond functionalization
  • Synthesis of [(triisopropylsilyl)ethynyl]indole derivatives
  • Domino condensation / S-arylation / heterocyclization reactions
Palladium-catalyzed carbonylation of iodoanilines provides ketoamides in the presence of primary and secondary amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 12051-12051 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service