Skip to Content
Merck
All Photos(1)

Documents

536717

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 6.0 solution

5 wt. % in methanol

Synonym(s):

Poly(amidoamine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=6);dendri PAMAM(NH2)256
CAS Number:
Molecular Weight:
58046.11
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

Quality Level

form

liquid

mol wt

generation 6.0

feature

core type ethylenediamine core (2-carbon core)

concentration

5 wt. % in methanol

no. Surface Groups

256

density

0.806 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 256 surface primary amino groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service