Skip to Content
Merck
All Photos(1)

Key Documents

522554

Sigma-Aldrich

2-Hydroxyphenylboronic acid pinacol ester

98%

Synonym(s):

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17BO3
CAS Number:
Molecular Weight:
220.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.506 (lit.)

bp

282 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccccc2O

InChI

1S/C12H17BO3/c1-11(2)12(3,4)16-13(15-11)9-7-5-6-8-10(9)14/h5-8,14H,1-4H3

InChI key

VLROJECCXBBKPZ-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of indolo-fused heterocyclic inhibitors of polymerase enzyme of hepatitis C
  • Studies of pi-interactions, electronic structure and transient UV absorption of subphthalocyanine-borate-bridged ferrocene-fullerene conjugates
  • Synthesis of subphthalocyanine and fused-ring nicotine derivatives
  • Suzuki-Miyaura coupling-triflation sequence, reduction and salt formation for synthesis of hydroxylated oligoarene phosphines
Widely used in Suzuki coupling chemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 58, 9633-9695 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service