Skip to Content
Merck
All Photos(3)

Documents

428841

Sigma-Aldrich

Martin Sulfurane Dehydrating agent

technical grade

Synonym(s):

Bis[α,α-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, Bis[α,α-bis(trifluoromethyl)benzyloxy]diphenylsulfur

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5C(CF3)2O]2S(C6H5)2
CAS Number:
Molecular Weight:
672.52
Beilstein:
1416879
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

mp

106-108 °C (lit.)

SMILES string

FC(F)(F)C(OS(OC(c1ccccc1)(C(F)(F)F)C(F)(F)F)(c2ccccc2)c3ccccc3)(c4ccccc4)C(F)(F)F

InChI

1S/C30H20F12O2S/c31-27(32,33)25(28(34,35)36,21-13-5-1-6-14-21)43-45(23-17-9-3-10-18-23,24-19-11-4-12-20-24)44-26(29(37,38)39,30(40,41)42)22-15-7-2-8-16-22/h1-20H

InChI key

RMIBJVUYNZSLSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile dehydrating and oxidizing agent. For chemistry and references, see Aldrichimica Acta .

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 81-81 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service