Merck
Studies on new, centrally active and reversible acetylcholinesterase inhibitors.
Neurochemical research (1990-06-01)
F Arnal, L J Coté, S Ginsburg, G D Lawrence, A Naini, M Sano
RÉSUMÉ
We have synthesized the tertiary amines of pyridostigmine and neostigmine, 3-pyridinol dimethylcarbamate (norpyridostigmine) and 3-dimethylaminophenol dimethylcarbamate (norneostigmine) respectively, and we have tested their abilities to cross the blood-brain barrier and inhibit mouse brain AChE activity. The in vivo inhibition of AChE activity by norpyridostigmine reaches 72% at 10 minutes which is comparable to that seen with physostigmine (73% at 10 minutes). Inhibition by norneostigmine is less effective (50% at 10 minutes) and approaches that obtained with tetrahydroaminoacridine (57% at 10 minutes). These data show that both norpyridostigmine and norneostigmine cross the blood-brain barrier and that they are effective inhibitors of mouse brain AChE activity. These drugs could be useful in the treatment of memory impairment associated with Alzheimer's disease, and other memory disorders.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Pyridostigmine impurity A, European Pharmacopoeia (EP) Reference Standard