Merck
Adriamycin promotes neurite outgrowth in the "neurite-minus" N1A-103 mouse neuroblastoma cell line.
Experimental cell research (1992-11-01)
J C Larcher, L Cordeau-Lossouarn, G Romey, F Gros, B Croizat, J L Vayssiere
RÉSUMÉ
Adriamycin, an anticancer agent acting on topoisomerase II, promotes the arrest of cell division and neurite extension in a "neurite-minus" murine neuroblastoma cell line, N1A-103. This morphological differentiation is accompanied by a blockade in the S phase of the cell cycle, modification of the amount of peripherin, and appearance of the beta 7-tubulin isoform. Yet, adriamycin-induced N1A-103 cells fail to express other neuronal markers, such as long-lasting Ca2+ channels, synaptophysin, and the shift in the proportion of the beta'1 tubulin isoform to the beta'2 isoform, whose appearance parallels the terminal differentiation of the wild type neuroblastoma cell line N1E-115. Hence, a comparison of the behavior of these two cell lines leads to the proposal that there are two programs of neuroblastoma differentiation: one where expression is triggered by the arrest of cell division and which is observed in adriamycin-induced N1A-103 variant cells, and the other, presumably occurring further downstream, which would involve further changes in morphogenesis and acquisition of new electrophysiological properties.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Methyl-1-cyclohexanecarboxylic acid, 99%