20638
Septa Thermogreen® LB-2, disques solides
diam. 5 mm ( 3/16 in.), pkg of 50 ea
Conditionnement
pkg of 50 ea
Diamètre
5 mm ( 3/16 in.)
Compatibilité
for use with Agilent/HP On-Column capillary port
Description générale
Septum is a very important component of syringe injection system. They are usually fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene or fluoroelastomers. It is suitable to conveniently introducing the sample without causing the system to leak. Improper septa decomposes under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
Version améliorée du septum Thermogreen LB-1 d′origine.
- Ressuage extrêmement faible sur une large gamme de températures d′injection (de 100 °C à 350 °C).
- Produit déjà conditionné, prêt à l'emploi.
- Pénétration de l′aiguille facilitée et grande tolérance aux piqûres (produit idéal pour les passeurs d′échantillons).
- Formulation de caoutchouc brevetée par Supelco.
Application
Thermogreen® LB-2 Septa is suitable for the following applications:
- To minimize extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS.
- Thermo-resistant septa is used in GC instrument to avoid carrier gas leaks, extraneous peaks and phthalate contamination.
- Thermo-resistant septa along with thick needle protector is used in GC instruments to avoid damage irreversibly caused by microsyringe.
- It is also used in the influent sampling ports in gas chromatographs.
Informations légales
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificats d'analyse (COA)
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Déjà en possession de ce produit ?
Solid-phase microextraction (SPME) for rapid field sampling and analysis by gas chromatography-mass spectrometry (GC-MS).
TrAC, Trends in Analytical Chemistry, 21, 534-543 (2002)
Journal of chromatography. A, 999(1-2), 145-153 (2003-07-30)
A simple and efficient liquid-phase microextraction (LPME) technique using a hollow-fibre membrane, in conjunction with gas chromatography-mass spectrometry has been developed for the extraction and analysis of six phthalate esters in water samples. Parameters such as extraction solvent, agitation of
Effect of periods of non-use on biofilter performance.
Journal of the Air & Waste Management Association (1995), 46, 539-546 (1996)
Modern Practice of Gas Chromatography, 471-472 (2004)
Chemosphere, 54(7), 849-857 (2003-11-26)
The sonochemical degradation of aqueous solutions containing low concentrations of six phthalate esters at an ultrasonic frequency of 80 kHz has been investigated. Ultrasonic treatment was found capable of removing the four higher molecular mass phthalates (di-n-butyl phthalate, butylbenzyl phthalate