Principaux documents
Z380067
Sigma-Aldrich
Brayer for Breathe-Easy® sealing membrane
4" soft rubber roller
Synonyme(s) :
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.22
Matériaux
rubber (soft)
Stérilité
non-sterile
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Diversified Biotech BRAY-4000
Description générale
Soft rubber roller (4") assures adhesion of Breathe-Easy membrane and facilitates removal of top protective sheet.
Informations légales
Breathe-Easy is a registered trademark of Diversified Biotech