Principaux documents
WHAWB100074
Unicore Punch Kit
I.D. 1.2 mm, pkg of 25 ea
Synonyme(s) :
UniCore Punches 1.20 mm (25), Whatman FTA, Z708828, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Qiagen WB100074
D.I.
1.2 mm
Catégories apparentées
Description générale
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Manual UniCore punch system for sample disc removal from FTA cards.
Caractéristiques et avantages
  • Stainless steel cutting tip (ranging from 1.2 mm) enables manual coring, retrieval, and controlled ejection of the FTA punch.
  • Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Autres remarques
Field of Use : For internal research use only
Informations légales
FTA is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Protocoles
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.