Principaux documents
TMO382019-0250
Nalgene® Certified Clean bottles
clear PETG, volume (250 mL), case of 48 ea
Synonyme(s) :
PETG bottle, certified nalgene bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
clear PETG
Conditionnement
case of 48 ea
Fabricant/nom de marque
Nalgene 382019-0250
Volume
(250 mL)
Description générale
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries