Merck
OH30456392
Ohaus® Navigator® Portable Balance
Model NVT2200, weighing capacity 2200 g, resolution: 1 g, AC/DC input 230 V, universal plug set
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.71
UNSPSC Code:
41111502
eCl@ss:
32120102
Feature:
CE compliant
Material:
metal weighing plate
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30456392
weighing capacity measuring range
2200 g, resolution: 1 g
weighing pan W × L
17.4 cm × 23 cm
General description
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Application
Weighing, Parts Counting, Percent Weighing, Checkweighing
Features and Benefits
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Legal Information
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.