Merck
MTCC3215SG
MTC™ Bio ClearSeal™ Graduated Screw Cap Microtubes
capacity 1.5 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea)
Synonyme(s) :
centrifuge tube, screw-cap tube, MTC Bio C3215-SG, sample collection tube, sample receptacle, sample tube
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.15
UNSPSC Code:
41121703
Capacity:
1.5 mL
Material:
clear , polypropylene , round , screw top cap
Sterility:
sterile
Packaging:
pack of 1000 ea (20 x bags 50 ea)
Parameter:
22,000 × g speed
Feature:
cap natural, graduation, neck thread external
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
clear , polypropylene , round , screw top cap
sterility
sterile
feature
cap natural, graduation, neck thread external
packaging
pack of 1000 ea (20 x bags 50 ea)
manufacturer/tradename
MTC™ Bio C3215-SG
parameter
22,000 × g speed
capacity
1.5 mL
diam. × L
12.8 mm × 47.6 mm
General description
ClearSeal™ screw-cap microtubes, 1.5mL , w/ O-ring & caps, sterile, printed graduations, self-standing, 20 resealable bags of 50 tubes, 1000/cs
Screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -86°C to +121°C and can be centrifuged at up to 22,000×g.
Features and Benefits
  • Ultra-clear polypropylene
  • Packaged in resealable bags of 50
  • Leak-proof O-ring seal
  • Sterile, RNase, DNase & Pyrogen free
  • White, printed graduations
Legal Information
ClearSeal is a trademark of MTC Bio
MTC is a trademark of MTC Bio