Principaux documents
BMSIPD96004M
Sigma-Aldrich
Benchmark Stainless Steel Bulk Beads
bead diam. 4 mm, acid washed
Synonyme(s) :
Stainless Steel Beads
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.82
Matériaux
stainless steel
Conditionnement
pack of 5000 ea
Fabricant/nom de marque
Benchmark Scientific IPD9600-4M
Diam. des billes
4 mm
Compatibilité
stainless steel grinding jars
Description générale
Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free