Principaux documents
Z743921
Ohaus® Explorer® precision balance
Model EX223/E, weighing capacity 220 g, Precision 1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Synonyme(s) :
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
Matériaux
stainless steel weighing plate
Caractéristiques
Model EX223/E
Fabricant/nom de marque
Ohaus® 30061979
Precision measuring range
1 mg
Plage de mesure de la capacité de pesée
220 g
Taille de la plateforme
5.1 in. (130 mm)
Description générale
Display
5.7" (145 mm) Full-color VGA graphic touchscreen display with user-controlled brightness
Operation
AC adapter (included)
Communication
RS232 and USB (included)
Construction
Modular design, metal base, ABS top housing, stainless steel pan, glass draftshield with top mounted side doors and flip/sliding top door (1mg models only), illuminated up-front level indicator, integral weigh below hook, security bracket, calibration lock, four touchless sensors, full housing in-use cover
Design Features
Selectable environmental filters, auto tare, user selectable span calibration points, software lockout and reset menu, user selectable communication settings and data print options, user definable project and user IDs, auto standby, up to 14 operating languages
Informations légales
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.