Key Documents
Z743165
Julabo® bath covers
flat cover to fit CORIO C-B5, CD-B5 and CD-BT5
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 9970296
Description générale
Stainless Steel Flat bath cover for 5L bath tanks. Suitable for use with Julabo® CORIO C-B5, CD-B5, CD-BT5.
Informations légales
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.