Principaux documents
Z740706
Sigma-Aldrich
Aldrich® Essentials boiling flask, round-bottom, ST/NS 24/40
capacity 50 mL
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass flask
round bottom
single-neck
Conditionnement
pkg of 1 ea
Capacité
50 mL
Description générale
Clear borosilicate glass, round bottomed boiling flasks with outer ST ground joint neck. White marking spot.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC