Principaux documents
Z682640
Ace evaporating flask, pear-shaped
capacity 2000 mL, joint: ST/NS 29/42, poly-coated, heavy wall
Synonyme(s) :
Aldrich® evaporating flask, pear-shaped, bottom flask, flask round, round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
VB.02
Matériaux
borosilicate glass
pear-shaped flask
single-neck
Joint
joint: ST/NS29/42
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Ace Glass 3990-124
Capacité
2000 mL
Description générale
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC