Principaux documents
Z682403
Ace glass steam tube, for condenser "C", "CR", "A" and "V" assemblies
fits Büchi® models R-200/R-205 and R-114/R-144
Synonyme(s) :
40610
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Büchi equivalent, 40610
Catégories apparentées
Description générale
The glass steam tube accessory fits the Büchi condenser assemblies, "C", "CR", "A" and "V", and works with Büchi Rotavapor models R-200/R-205 and R-114/R-144.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG