Principaux documents
Z549576
Sigma-Aldrich
Stopcock Adapter
glass side-port stopcock, joint: ST/NS 19/22
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Matériaux
glass side-port stopcock
Joint
joint: ST/NS19/22
Description générale
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.