Key Documents
Z531235
Sigma-Aldrich
Aldrich® coiled condenser
jacketed, L 200 mm, joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
Longueur
200 mm
Description générale
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC