Key Documents
Z530360
Sigma-Aldrich
Aldrich® in-line oil bubbler
joint: ST/NS 19/22
Synonyme(s) :
bubblers, in-line bubblers, scientific bubblers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS19/22
Capacité
10 mL
Catégories apparentées
Description générale
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC