Principaux documents
Z503231
Sigma-Aldrich
Funnel holder
Double
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Matériaux
high-density polyethylene
Caractéristiques
Holds 1 x 25 to 152 mm diameter funnel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18242-0000
Description générale
Holds 76 to 152 mm diameter funnels. Comes with removable inner ring that can hold smaller funnels between 25 to 76 mm.