Principaux documents
Z408735
Ace gas dispersion tube with porous fritted glass tip
Fit #11 Ace-Thred, O.D. × L 10 mm × 150 mm, porosity 70-100 μm (B)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Fabricant/nom de marque
Ace Glass 720204
D.E. × L
10 mm × 150 mm
Dimension de pores
70-100 μm porosity (B)