Principaux documents
Z300969
Quickfit® Reduction Adapters
top female joint: ST/NS 24/29, bottom male joint: ST/NS 29/32
Synonyme(s) :
DA34
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS top24/29
male joint: ST/NS bottom29/32
Fabricant/nom de marque
Quickfit DA 34
Disponibilité
not available in USA and Canada
Description générale
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited