Z285501
DigiTrol II temperature controller
AC/DC input 240 V AC, 10 A (CE Compliant)
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.76
Produits recommandés
Fabricant/nom de marque
Glas-Col 104APL624K (CE)
Paramètres
32-999 °F temperature (0-750 °C)
Sortie
2,400 watts
Description générale
Simultaneously displays set point and bath temperature to 1.0°C. Supplied with a Type K, 6 in. immersion thermocouple probe that fits Aldrich oil baths. 240 V models are supplied with a US connector.