Principaux documents
Z282081
Sigma-Aldrich
Coors apothecary mortar
O.D. × H 100 mm × 59 mm
About This Item
Code UNSPSC :
41101702
Nomenclature NACRES :
NB.35
Matériaux
alumina
Fabricant/nom de marque
Coorstek 60382
D.E. × H
100 mm × 59 mm
Description générale
99.5% alumina. Extremely hard and completely nonabsorbent surface. Mortar shaped for easy handling. Outside glazed except for bottom. Order pestle separately.
Informations légales
Coors is a trademark of Coorstek, Inc.
Les clients ont également consulté
Slide 1 of 1