Key Documents
Z261297
Stainless steel 316 syringe needle, noncoring point
gauge 20, L 8 in.
Synonyme(s) :
Stainless steel needle
About This Item
Code UNSPSC :
42142600
Nomenclature NACRES :
NB.03
Description
Micro-Mate Hub with 316 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Popper 9864
Longueur
8 in.
Calibre
20
Description générale
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing