Principaux documents
Z232386
Pouring rings for Duran® laboratory bottles
GL 32 neck thread, red melamine resin
Synonyme(s) :
DURAN pouring ring, ETFE, GL32, high temperature, pouring ring
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux:
red melamine resin
Raccords:
GL 32 neck thread
Matériaux
red melamine resin
Fabricant/nom de marque
DURAN®
Schott
Paramètres
-100-180 °C temp. stability
Raccords
GL 32 neck thread
Description générale
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
Caractéristiques et avantages
Optional pouring rings prevent drips when pouring from Duran bottles.
Informations légales
DURAN is a registered trademark of DWK Life Sciences