Key Documents
Z222380
Metal standard taper clip
joint size, 24
Synonyme(s) :
Metal clip, Taper clip
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Description générale
Nickel-plated spring-steel joint clips offer increased strength and resistance to heat and cracking over Delrin® plastic type. Securely holds conical joints under pressure at temperatures up to 392 °F (200 °C). Will not scratch glassware.
Informations légales
Delrin is a registered trademark of E. I. du Pont de Nemours and Company